Product Details

SNP ID
rs11842971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:76882685 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCAACACTGACTTTTGGTCAGT[C/G]AATCAGAGTCCTAAATTATTTCAGG
Phenotype
MIM: 610521
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KCTD12 PubMed Links

Gene Details

Gene
KCTD12
Gene Name
potassium channel tetramerization domain containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138444.3 3721 UTR 3 NP_612453.1

View Full Product Details