Product Details
- SNP ID
-
rs1294087
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:154016460 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTAATACATATACATATCTGTTTA[C/T]GGTAACAATTTATGGAGAAAACAAA
- Phenotype
-
MIM: 600018
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OPRM1
PubMed Links
Gene Details
- Gene
- OPRM1
- Gene Name
- opioid receptor mu 1
View Full Product Details