Product Details

SNP ID
rs35448589
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:79844395 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAACAAGACCTGGTGACAGGATG[A/G]ACATGGAGTGTGAGTAAGAGAGCCA
Phenotype
MIM: 604197
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MTHFS PubMed Links

Gene Details

Gene
MTHFS
Gene Name
5,10-methenyltetrahydrofolate synthetase (5-formyltetrahydrofolate cyclo-ligase)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199758.1 1434 UTR 3 NP_001186687.1
NM_006441.3 1434 UTR 3 NP_006432.1
Gene
ST20-MTHFS
Gene Name
ST20-MTHFS readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199760.1 1434 UTR 3 NP_001186689.1

View Full Product Details