Product Details

SNP ID
rs220196
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.21:42014253 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATGCCCCTTGGCACAGAGACCATGT[C/T]TGTTAACTTGTGTATGCCCGGAGCT
Phenotype
MIM: 616485
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ZBTB21 PubMed Links

Gene Details

Gene
ZBTB21
Gene Name
zinc finger and BTB domain containing 21
There are no transcripts associated with this gene.

Gene
ZNF295-AS1
Gene Name
ZNF295 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details