Product Details
- SNP ID
-
rs7014582
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:143916360 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CGGTGCGCGAGCCGGTGCGGGAGCC[A/G]GCGGTAGAGCCGGAGCCGCTGACGC
- Phenotype
-
MIM: 601282
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PLEC
PubMed Links
Gene Details
- Gene
- PLEC
- Gene Name
- plectin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000445.4 |
13407 |
Silent Mutation |
GCC,GCT |
A4514A |
NP_000436.2 |
NM_201378.3 |
13407 |
Silent Mutation |
GCC,GCT |
A4473A |
NP_958780.1 |
NM_201379.2 |
13407 |
Silent Mutation |
GCC,GCT |
A4465A |
NP_958781.1 |
NM_201380.3 |
13407 |
Silent Mutation |
GCC,GCT |
A4624A |
NP_958782.1 |
NM_201381.2 |
13407 |
Silent Mutation |
GCC,GCT |
A4455A |
NP_958783.1 |
NM_201382.3 |
13407 |
Silent Mutation |
GCC,GCT |
A4487A |
NP_958784.1 |
NM_201383.2 |
13407 |
Silent Mutation |
GCC,GCT |
A4491A |
NP_958785.1 |
NM_201384.2 |
13407 |
Silent Mutation |
GCC,GCT |
A4487A |
NP_958786.1 |
XM_005250976.3 |
13407 |
Silent Mutation |
GCC,GCT |
A4629A |
XP_005251033.1 |
XM_005250978.3 |
13407 |
Silent Mutation |
GCC,GCT |
A4496A |
XP_005251035.1 |
XM_005250979.4 |
13407 |
Silent Mutation |
GCC,GCT |
A4492A |
XP_005251036.1 |
XM_005250980.4 |
13407 |
Silent Mutation |
GCC,GCT |
A4492A |
XP_005251037.1 |
XM_005250981.3 |
13407 |
Silent Mutation |
GCC,GCT |
A4478A |
XP_005251038.1 |
XM_005250982.3 |
13407 |
Silent Mutation |
GCC,GCT |
A4470A |
XP_005251039.1 |
XM_005250983.2 |
13407 |
Silent Mutation |
GCC,GCT |
A4464A |
XP_005251040.1 |
XM_005250984.4 |
13407 |
Silent Mutation |
GCC,GCT |
A4460A |
XP_005251041.1 |
XM_006716588.3 |
13407 |
Silent Mutation |
GCC,GCT |
A4519A |
XP_006716651.1 |
XM_006716589.2 |
13407 |
Silent Mutation |
GCC,GCT |
A4469A |
XP_006716652.1 |
XM_006716590.3 |
13407 |
Silent Mutation |
GCC,GCT |
A4469A |
XP_006716653.1 |
XM_011517130.2 |
13407 |
Silent Mutation |
GCC,GCT |
A4492A |
XP_011515432.1 |
XM_011517131.2 |
13407 |
Silent Mutation |
GCC,GCT |
A4464A |
XP_011515433.1 |
XM_011517132.2 |
13407 |
Silent Mutation |
GCC,GCT |
A3369A |
XP_011515434.1 |
View Full Product Details