Product Details

SNP ID
rs2075650
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44892362 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGATGAGAGTTGGTGTGGGGTTGG[A/G]GTGGAGTGTGACAGCGTTTCTCTTC
Phenotype
MIM: 600798 MIM: 608061
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NECTIN2 PubMed Links

Gene Details

Gene
NECTIN2
Gene Name
nectin cell adhesion molecule 2
There are no transcripts associated with this gene.

Gene
TOMM40
Gene Name
translocase of outer mitochondrial membrane 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128916.1 Intron NP_001122388.1
NM_001128917.1 Intron NP_001122389.1
NM_006114.2 Intron NP_006105.1
XM_005258411.3 Intron XP_005258468.1

View Full Product Details