Product Details

SNP ID
rs11227247
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:65655382 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCTTGTCAGTTTTTGAGCCTCGCAC[C/A]GGCACACACATCTTGCTAATTTAGA
Phenotype
MIM: 164014 MIM: 602180
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
MIR4489 PubMed Links

Gene Details

Gene
MIR4489
Gene Name
microRNA 4489
There are no transcripts associated with this gene.

Gene
RELA
Gene Name
RELA proto-oncogene, NF-kB subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145138.1 Intron NP_001138610.1
NM_001243984.1 Intron NP_001230913.1
NM_001243985.1 Intron NP_001230914.1
NM_021975.3 Intron NP_068810.3
XM_011545206.1 Intron XP_011543508.1
XM_011545207.1 Intron XP_011543509.1
Gene
SIPA1
Gene Name
signal-induced proliferation-associated 1
There are no transcripts associated with this gene.

View Full Product Details