Product Details

SNP ID
rs35023929
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:56634201 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CCTTGCGGGAGAAGTTGGACAAGAT[C/T]GGGTTGAATCTTCCGGCCGGGAGGC
Phenotype
MIM: 601602
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TFAP2C PubMed Links

Gene Details

Gene
TFAP2C
Gene Name
transcription factor AP-2 gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003222.3 1098 Silent Mutation ATC,ATT I285I NP_003213.1

View Full Product Details