Product Details

SNP ID
rs339694
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:42696387 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAACCACCTGCCCACACTCAGGA[C/T]GCTCTGAGTTCCATCAGTCCTCTGG
Phenotype
MIM: 608116 MIM: 615340
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC13 PubMed Links

Gene Details

Gene
CCDC13
Gene Name
coiled-coil domain containing 13
There are no transcripts associated with this gene.

Gene
HHATL
Gene Name
hedgehog acyltransferase-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020707.3 Intron NP_065758.3
XM_006713274.2 Intron XP_006713337.1
XM_006713275.2 Intron XP_006713338.1
XM_011533969.2 Intron XP_011532271.1
XM_011533970.2 Intron XP_011532272.1
XM_017006935.1 Intron XP_016862424.1
XM_017006936.1 Intron XP_016862425.1
XM_017006937.1 Intron XP_016862426.1
XM_017006938.1 Intron XP_016862427.1
Gene
KLHL40
Gene Name
kelch like family member 40
There are no transcripts associated with this gene.

View Full Product Details