Product Details
- SNP ID
-
rs2287937
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:8065536 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TCTACTCCGAGGAATAAGCCCTGTG[C/G]CCACCCCAGTTGCCCATTGCCATGT
- Phenotype
-
MIM: 602565
MIM: 608529
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CCL25
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77141825] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCL25
- Gene Name
- C-C motif chemokine ligand 25
There are no transcripts associated with this gene.
- Gene
- FBN3
- Gene Name
- fibrillin 3
View Full Product Details