Product Details
- SNP ID
-
rs1478604
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:39581120 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGGCCTGCGAGTTCAGGGCTCCTG[C/T]CGCTCTCCAGGAGCAACCTCTACTC
- Phenotype
-
MIM: 188060
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
THBS1
PubMed Links
Gene Details
- Gene
- THBS1
- Gene Name
- thrombospondin 1
View Full Product Details