Product Details

SNP ID
rs553485
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:111259531 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CGGCAGCAAGACGACTTCTCCGGAG[C/T]CGCCGAGCTGGAGTTAGAGGTGGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MFSD4B PubMed Links

Gene Details

Gene
MFSD4B
Gene Name
major facilitator superfamily domain containing 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153369.2 253 UTR 5 NP_699200.2
XM_017011483.1 253 UTR 5 XP_016866972.1

View Full Product Details