Product Details
- SNP ID
-
rs309248
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:68715237 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTAAGAATCCTTTGCGCTCTCGG[C/T]GCCCCCCTCCACGCCCAGGAGCGTG
- Phenotype
-
MIM: 616102
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC102B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs199817898] are located under a probe and SNP(s) [rs74917552] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCDC102B
- Gene Name
- coiled-coil domain containing 102B
- Gene
- TMX3
- Gene Name
- thioredoxin related transmembrane protein 3
View Full Product Details