Product Details

SNP ID
rs3804465
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:6591071 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCAGTGCAGGATGAGAAGCAAGATG[G/T]GAGATAAATGGACTGGTCAGGGTTG
Phenotype
MIM: 605241
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LY86 PubMed Links

Gene Details

Gene
LY86
Gene Name
lymphocyte antigen 86
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004271.3 Intron NP_004262.1
XM_017011505.1 Intron XP_016866994.1
Gene
LY86-AS1
Gene Name
LY86 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details