Product Details
- SNP ID
-
rs11032700
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:34440156 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGCAGCCCTGTTTTCCCTATTCTG[A/C]CTAGTGCTCTTGCTCAAAACTCAGG
- Phenotype
-
MIM: 115500
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CAT
PubMed Links
Gene Details
- Gene
- CAT
- Gene Name
- catalase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001752.3 |
|
Intron |
|
|
NP_001743.1 |
View Full Product Details