Product Details

SNP ID
rs11236733
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:76359878 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATAAAGGTCCACAAATAGTTAAG[A/G]AAAAGAAGAGTTAAACAATCACCTT
Phenotype
MIM: 607374
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
THAP12 PubMed Links

Gene Details

Gene
THAP12
Gene Name
THAP domain containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004705.3 Intron NP_004696.2

View Full Product Details