Product Details

SNP ID
rs11570176
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57761957 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGAGGAAGAGGTGGCGAAAAATCA[A/G]CTGCCCTGCTGGATTTGTCTTTCTC
Phenotype
MIM: 601045
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BTBD18 PubMed Links

Gene Details

Gene
BTBD18
Gene Name
BTB domain containing 18
There are no transcripts associated with this gene.

Gene
CTNND1
Gene Name
catenin delta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085458.1 196 UTR 5 NP_001078927.1
NM_001085459.1 196 UTR 5 NP_001078928.1
NM_001085460.1 196 UTR 5 NP_001078929.1
NM_001085461.1 196 UTR 5 NP_001078930.1
NM_001085462.1 196 UTR 5 NP_001078931.1
NM_001085463.1 196 UTR 5 NP_001078932.1
NM_001085464.1 196 UTR 5 NP_001078933.1
NM_001085465.1 196 UTR 5 NP_001078934.1
NM_001085466.1 196 UTR 5 NP_001078935.1
NM_001085467.1 196 UTR 5 NP_001078936.1
NM_001085468.1 196 UTR 5 NP_001078937.1
NM_001085469.1 196 UTR 5 NP_001078938.1
NM_001206883.1 196 UTR 5 NP_001193812.1
NM_001206884.1 196 UTR 5 NP_001193813.1
NM_001206885.1 196 UTR 5 NP_001193814.1
NM_001206886.1 196 UTR 5 NP_001193815.1
NM_001206887.1 196 UTR 5 NP_001193816.1
NM_001206888.1 196 UTR 5 NP_001193817.1
NM_001206889.1 196 UTR 5 NP_001193818.1
NM_001206890.1 196 UTR 5 NP_001193819.1
NM_001206891.1 196 UTR 5 NP_001193820.1
NM_001331.2 196 UTR 5 NP_001322.1
Gene
TMX2-CTNND1
Gene Name
TMX2-CTNND1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details