Product Details
- SNP ID
-
rs35729071
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:35713525 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTCTTTCTGTCTCCTCCCTGCCAAC[C/T]TAGAGCCCTCTCCCTGAGGTGTCCA
- Phenotype
-
MIM: 606834
MIM: 605859
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KMT2B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75903850] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KMT2B
- Gene Name
- lysine methyltransferase 2B
There are no transcripts associated with this gene.
- Gene
- ZBTB32
- Gene Name
- zinc finger and BTB domain containing 32
View Full Product Details