Product Details

SNP ID
rs34789088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:50995242 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAAATGGAGAAATAACACCTGCC[C/T]TCCAGGGTTGTTATGATCATTAAAC
Phenotype
MIM: 603695
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GUCY1B2 PubMed Links

Gene Details

Gene
GUCY1B2
Gene Name
guanylate cyclase 1 soluble subunit beta 2 (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details