Product Details
- SNP ID
-
rs10509407
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:87503230 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGTACTGTGCCATACATTTTGTAAG[C/T]GGAGTCTCTAAAGATTGCTTCATAA
- Phenotype
-
MIM: 605391
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MINPP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142349941] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MINPP1
- Gene Name
- multiple inositol-polyphosphate phosphatase 1
- Gene
- MIR4678
- Gene Name
- microRNA 4678
There are no transcripts associated with this gene.
View Full Product Details