Product Details

SNP ID
rs11841182
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.13:20187704 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TTCAATCTTCCTATAGATCTGCTCA[A/T]TATTTATCTAAACCTTAGCTTCTAT
Phenotype
MIM: 121011
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GJB2 PubMed Links

Gene Details

Gene
GJB2
Gene Name
gap junction protein beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004004.5 2084 UTR 3 NP_003995.2
XM_011535049.2 2084 UTR 3 XP_011533351.1

View Full Product Details