Product Details

SNP ID
rs1980749
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:3721767 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACTTATGGCTCTTTCAAGTGTGG[A/G]AATATAAAAAATGAACTTATGAGAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100507336 PubMed Links

Gene Details

Gene
LOC100507336
Gene Name
uncharacterized LOC100507336
There are no transcripts associated with this gene.

Gene
PXDC1
Gene Name
PX domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_183373.3 Intron NP_899229.2
XM_011514393.2 Intron XP_011512695.1

View Full Product Details