Product Details
- SNP ID
-
rs272827
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
69
- Location
-
Chr.1:36161590 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGTTAAATGAACTAATGAGTTTAA[C/T]GTGTTTAGCTGTGCCTGACAAAAGC
- Phenotype
-
MIM: 610955
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MAP7D1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79152495] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MAP7D1
- Gene Name
- MAP7 domain containing 1
- Gene
- TRAPPC3
- Gene Name
- trafficking protein particle complex 3
There are no transcripts associated with this gene.
View Full Product Details