Product Details
- SNP ID
-
rs4912389
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
44
- Location
-
Chr.1:59138085 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTCCTTGGCTCCCCCAACCCCCAC[A/G]CATCCTGTCACATCCCAGATCAGAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HSD52
PubMed Links
Gene Details
- Gene
- HSD52
- Gene Name
- uncharacterized LOC729467
There are no transcripts associated with this gene.
View Full Product Details