Product Details

SNP ID
rs7956271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52602114 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGAGGGTTGACTTTGGACCAGAG[C/T]GAAAAGATTTCTCCACTAAGCATTT
Phenotype
MIM: 608246 MIM: 608247
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT72 PubMed Links

Gene Details

Gene
KRT72
Gene Name
keratin 72
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146225.1 Intron NP_001139697.1
NM_001146226.1 Intron NP_001139698.1
NM_080747.2 Intron NP_542785.1
XM_017018833.1 Intron XP_016874322.1
XM_017018834.1 Intron XP_016874323.1
XM_017018835.1 Intron XP_016874324.1
XM_017018836.1 Intron XP_016874325.1
XM_017018837.1 Intron XP_016874326.1
Gene
KRT73
Gene Name
keratin 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175068.2 Intron NP_778238.1
XM_011538257.2 Intron XP_011536559.1
Gene
KRT73-AS1
Gene Name
KRT73 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details