Product Details

SNP ID
rs932936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:22897690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCCTCGCCGCCATTAGCAGGGA[C/G]GGGGGCTGCCTGCTCTTTCTCGGGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BASP1P1 PubMed Links

Gene Details

Gene
BASP1P1
Gene Name
brain abundant, membrane attached signal protein 1 pseudogene 1
There are no transcripts associated with this gene.

Gene
LINC00621
Gene Name
long intergenic non-protein coding RNA 621
There are no transcripts associated with this gene.

Gene
LOC105370109
Gene Name
uncharacterized LOC105370109
There are no transcripts associated with this gene.

View Full Product Details