Product Details
- SNP ID
-
rs11610085
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:25960762 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- TTCTTTCTAGGTTTATTGATCAAAT[A/C]TTATGTCTGGGCTTTAGTAGCTTAG
- Phenotype
-
MIM: 608231
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
RASSF8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs61914182] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RASSF8
- Gene Name
- Ras association domain family member 8
- Gene
- RASSF8-AS1
- Gene Name
- RASSF8 atnisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details