Product Details

SNP ID
rs26742
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:16664660 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCAAACTGCCTACCTGTGTGTTCT[A/G]GTCCACAGGGAGGTGCAGGCAGGTT
Phenotype
MIM: 601481
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYO10 PubMed Links

Gene Details

Gene
MYO10
Gene Name
myosin X
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012334.2 8677 UTR 3 NP_036466.2
XM_005248306.4 8677 Intron XP_005248363.1
XM_005248307.2 8677 Intron XP_005248364.1
XM_006714475.2 8677 Intron XP_006714538.1
XM_011514046.2 8677 Intron XP_011512348.1

View Full Product Details