Product Details
- SNP ID
-
rs1227236
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:64826548 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCAAGACACTAAGAACGACTACCCA[A/G]AAAGCACTGCTGTACCTGTGTGGTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANXA2P3
PubMed Links
Gene Details
- Gene
- ANXA2P3
- Gene Name
- annexin A2 pseudogene 3
There are no transcripts associated with this gene.
View Full Product Details