Product Details

SNP ID
rs11785273
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:132782235 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGTTATATTGGAGACACTGTAAG[C/T]GGGTGGGTTTAAATTCAGACTGTTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PHF20L1 PubMed Links

Gene Details

Gene
PHF20L1
Gene Name
PHD finger protein 20-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277196.1 Intron NP_001264125.1
NM_016018.4 Intron NP_057102.4
NM_198513.1 Intron NP_940915.1
XM_005250931.1 Intron XP_005250988.1
XM_011517075.1 Intron XP_011515377.1
XM_011517076.1 Intron XP_011515378.1
XM_011517077.1 Intron XP_011515379.1
XM_011517078.1 Intron XP_011515380.1
XM_011517079.1 Intron XP_011515381.1
XM_011517080.1 Intron XP_011515382.1
XM_011517081.1 Intron XP_011515383.1
XM_011517088.1 Intron XP_011515390.1
XM_017013509.1 Intron XP_016868998.1
XM_017013510.1 Intron XP_016868999.1
XM_017013511.1 Intron XP_016869000.1
XM_017013512.1 Intron XP_016869001.1
XM_017013513.1 Intron XP_016869002.1
XM_017013514.1 Intron XP_016869003.1
XM_017013515.1 Intron XP_016869004.1
XM_017013516.1 Intron XP_016869005.1
XM_017013517.1 Intron XP_016869006.1

View Full Product Details