Product Details

SNP ID
rs2985700
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:155264144 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCTTGGGAGAGCCACTTCACTGT[C/T]GGGTAAAACAACGGAGCAAGATGAG
Phenotype
MIM: 607033 MIM: 604709
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLDN20 PubMed Links

Gene Details

Gene
CLDN20
Gene Name
claudin 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001346.3 132 UTR 5 NP_001001346.1
XM_011535848.2 132 Intron XP_011534150.1
Gene
TFB1M
Gene Name
transcription factor B1, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016020.3 132 Intron NP_057104.2
XM_005267006.4 132 Intron XP_005267063.1
XM_011535870.1 132 Intron XP_011534172.1
XM_011535871.1 132 Intron XP_011534173.1
XM_011535872.2 132 Intron XP_011534174.1
XM_011535873.2 132 Intron XP_011534175.1
XM_017010917.1 132 Intron XP_016866406.1
Gene
TIAM2
Gene Name
T-cell lymphoma invasion and metastasis 2
There are no transcripts associated with this gene.

View Full Product Details