Product Details

SNP ID
rs1749409
Assay Type
Validated
NCBI dbSNP Submissions
78
Location
Chr.1:155900625 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATCCTTCTTCCTTGGTGACCTTTAA[A/G]GGCAAAATGTGAGAAAAGATAACAG
Phenotype
MIM: 609591
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RIT1 PubMed Links

Gene Details

Gene
RIT1
Gene Name
Ras like without CAAX 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256820.1 Intron NP_001243749.1
NM_001256821.1 Intron NP_001243750.1
NM_006912.5 Intron NP_008843.1

View Full Product Details