Product Details

SNP ID
rs35653612
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:43144550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCTCATCATCCAGCCCTCCTTT[A/C]TGCTGTTTTTCCTCAGTTTTCAAGC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM183A PubMed Links

Gene Details

Gene
FAM183A
Gene Name
family with sequence similarity 183 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101376.2 1592 Intron NP_001094846.2
XM_005270875.4 1592 Intron XP_005270932.1
XM_005270876.4 1592 UTR 5 XP_005270933.1
XM_017001322.1 1592 UTR 5 XP_016856811.1

View Full Product Details