Product Details

SNP ID
rs511875
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:43145782 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGGACATACAAAGGGCCAAGAAG[C/T]GGCAAGACTGTCTTGAGAAGACATT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM183A PubMed Links

Gene Details

Gene
FAM183A
Gene Name
family with sequence similarity 183 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101376.2 Intron NP_001094846.2
XM_005270875.4 Intron XP_005270932.1
XM_005270876.4 Intron XP_005270933.1
XM_017001322.1 Intron XP_016856811.1

View Full Product Details