Product Details

SNP ID
rs1427198
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:58515210 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAAATTGCTTTGCAGTAATAGTT[G/T]CTTTTACAGAAAGTCTTAACATCTC
Phenotype
MIM: 614295
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BICC1 PubMed Links

Gene Details

Gene
BICC1
Gene Name
BicC family RNA binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080512.2 Intron NP_001073981.1
XM_005270169.4 Intron XP_005270226.1
XM_011540185.2 Intron XP_011538487.1
XM_011540188.2 Intron XP_011538490.2
XM_011540189.2 Intron XP_011538491.1
XM_011540190.2 Intron XP_011538492.1
XM_011540191.2 Intron XP_011538493.1
XM_017016677.1 Intron XP_016872166.1
XM_017016678.1 Intron XP_016872167.1

View Full Product Details