Product Details
- SNP ID
-
rs35776966
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:133987587 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTAAAATCTATGCAAGCTTAAGACT[C/T]TGGCTAAAGTGTGTTGGCTTCTTTT
- Phenotype
-
MIM: 610372
MIM: 605521
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SLC2A12
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142078499] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLC2A12
- Gene Name
- solute carrier family 2 member 12
- Gene
- TBPL1
- Gene Name
- TATA-box binding protein like 1
View Full Product Details