Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_138805.2 | Intron | NP_620160.1 | |||
XM_005264862.4 | Intron | XP_005264919.1 | |||
XM_005264863.4 | Intron | XP_005264920.1 | |||
XM_005264864.4 | Intron | XP_005264921.1 | |||
XM_006712965.3 | Intron | XP_006713028.1 | |||
XM_006712967.3 | Intron | XP_006713030.1 | |||
XM_011533348.2 | Intron | XP_011531650.1 | |||
XM_011533349.2 | Intron | XP_011531651.1 | |||
XM_011533350.2 | Intron | XP_011531652.1 | |||
XM_011533351.2 | Intron | XP_011531653.1 | |||
XM_011533352.2 | Intron | XP_011531654.1 | |||
XM_011533353.2 | Intron | XP_011531655.1 | |||
XM_011533354.2 | Intron | XP_011531656.1 |