Product Details

SNP ID
rs635901
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.18:32264099 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AACATGATGTAACCATGAGAATCCC[A/G]TATTATTACAATCCCACAATCTAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GAREM1 PubMed Links

Gene Details

Gene
GAREM1
Gene Name
GRB2 associated regulator of MAPK1 subtype 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242409.1 6456 UTR 3 NP_001229338.1
NM_022751.2 6456 UTR 3 NP_073588.1
XM_017025919.1 6456 Intron XP_016881408.1

View Full Product Details