Product Details

SNP ID
rs1561800
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:3803687 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTAATTCAGGATGAAATTATATGG[A/G]TGAAGTCAGAAAAACATTTATTCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100130207 PubMed Links

Gene Details

Gene
LOC100130207
Gene Name
uncharacterized LOC100130207
There are no transcripts associated with this gene.

Gene
LRRN1
Gene Name
leucine rich repeat neuronal 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324188.1 Intron NP_001311117.1
NM_001324189.1 Intron NP_001311118.1
NM_020873.6 Intron NP_065924.3

View Full Product Details