Product Details

SNP ID
rs1055256
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:124758023 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCCTCAATTCCCTTTGGACTTGCAC[A/G]CGCACTTCCTACACACAGAAGTGGC
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
METTL10 PubMed Links

Gene Details

Gene
METTL10
Gene Name
methyltransferase like 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304467.1 3468 UTR 3 NP_001291396.1
NM_001304468.1 3468 UTR 3 NP_001291397.1
NM_212554.3 3468 UTR 3 NP_997719.2

View Full Product Details