Product Details

SNP ID
rs37976
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:7975326 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTGTTTATTCCAAGTGAATGTTA[C/T]TAAGGCACATACCAAGTTCATGAAT
Phenotype
MIM: 614283
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLCCI1 PubMed Links

Gene Details

Gene
GLCCI1
Gene Name
glucocorticoid induced 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138426.3 Intron NP_612435.1
Gene
LOC100505921
Gene Name
uncharacterized LOC100505921
There are no transcripts associated with this gene.

View Full Product Details