Product Details

SNP ID
rs11048778
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:26939193 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTATCCGAACTAGAAAACCAGGA[A/G]TCATCATTGGTTCTTCATTTCCACT
Phenotype
MIM: 615079 MIM: 608858
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASUN PubMed Links

Gene Details

Gene
ASUN
Gene Name
asunder, spermatogenesis regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018164.2 Intron NP_060634.2
XM_005253422.3 Intron XP_005253479.1
XM_011520750.2 Intron XP_011519052.1
XM_011520751.2 Intron XP_011519053.1
XM_011520752.2 Intron XP_011519054.1
XM_011520753.2 Intron XP_011519055.1
XM_017019632.1 Intron XP_016875121.1
XM_017019633.1 Intron XP_016875122.1
XM_017019634.1 Intron XP_016875123.1
XM_017019635.1 Intron XP_016875124.1
XM_017019636.1 Intron XP_016875125.1
XM_017019637.1 Intron XP_016875126.1
Gene
FGFR1OP2
Gene Name
FGFR1 oncogene partner 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171887.1 Intron NP_001165358.1
NM_001171888.1 Intron NP_001165359.1
NM_015633.2 Intron NP_056448.1

View Full Product Details