Product Details
- SNP ID
-
rs11594574
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:17952237 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TACATGCCTTAAGCCATCAACAGAA[A/G]GATAATTTTTTTTTCCAGGAAGTTG
- Phenotype
-
MIM: 608734
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SLC39A12
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74642760] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLC39A12
- Gene Name
- solute carrier family 39 member 12
View Full Product Details