Product Details

SNP ID
rs12386036
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:21161816 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATATGATGTGAGTCACGTATGTAA[C/T]TTAGAATTTTTCTAGTAGCCACATG
Phenotype
MIM: 603334 MIM: 611304
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAH3 PubMed Links

Gene Details

Gene
DNAH3
Gene Name
dynein axonemal heavy chain 3
There are no transcripts associated with this gene.

Gene
TMEM159
Gene Name
transmembrane protein 159
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301769.1 Intron NP_001288698.1
NM_001301771.1 Intron NP_001288700.1
NM_001301773.1 Intron NP_001288702.1
NM_001301774.1 Intron NP_001288703.1
NM_001301775.1 Intron NP_001288704.1
NM_020422.5 Intron NP_065155.3
XM_005255439.4 Intron XP_005255496.1
XM_006721066.2 Intron XP_006721129.1
XM_006721067.3 Intron XP_006721130.1
XM_006721069.3 Intron XP_006721132.1
XM_017023471.1 Intron XP_016878960.1
XM_017023472.1 Intron XP_016878961.1
XM_017023473.1 Intron XP_016878962.1
XM_017023474.1 Intron XP_016878963.1

View Full Product Details