Product Details

SNP ID
rs11951903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:170107104 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGACACCCCTGCAGGGTAGCTCT[C/T]ATGAAACCCATTCTACAGATTAGGA
Phenotype
MIM: 601093
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FOXI1 PubMed Links

Gene Details

Gene
FOXI1
Gene Name
forkhead box I1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012188.4 Intron NP_036320.2
NM_144769.2 Intron NP_658982.1

View Full Product Details