Product Details

SNP ID
rs1469087
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:58061611 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTGACGTTTGAGGACGTGGTAGTGG[A/G]CTTCAGCCAGGAGGAGTGGGGGCAG
Phenotype
MIM: 604077
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF135 PubMed Links

Gene Details

Gene
ZNF135
Gene Name
zinc finger protein 135
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164527.1 104 Intron NP_001157999.1
NM_001164529.1 104 Missense Mutation GAC,GGC D34G NP_001158001.1
NM_001164530.1 104 Missense Mutation GAC,GGC D34G NP_001158002.1
NM_001289401.1 104 Missense Mutation GAC,GGC D22G NP_001276330.1
NM_001289402.1 104 UTR 5 NP_001276331.1
NM_003436.3 104 Missense Mutation GAC,GGC D22G NP_003427.3
NM_007134.1 104 Missense Mutation GAC,GGC D34G NP_009065.1
XM_005259211.1 104 Missense Mutation GAC,GGC D34G XP_005259268.1
XM_006723362.3 104 Missense Mutation GAC,GGC D22G XP_006723425.1
XM_006723363.3 104 Missense Mutation GAC,GGC D22G XP_006723426.1
XM_017027240.1 104 Missense Mutation GAC,GGC D22G XP_016882729.1
XM_017027241.1 104 UTR 5 XP_016882730.1
XM_017027242.1 104 Intron XP_016882731.1
Gene
ZSCAN1
Gene Name
zinc finger and SCAN domain containing 1
There are no transcripts associated with this gene.

View Full Product Details