Product Details

SNP ID
rs2270942
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48934744 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATCTGATCTCTCTTTCTTTTCCCC[A/G]CACAAACCTGAGTGTCTTGAGGTTT
Phenotype
MIM: 606377
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHDH PubMed Links

Gene Details

Gene
DHDH
Gene Name
dihydrodiol dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014475.3 Intron NP_055290.1
XM_005258748.4 Intron XP_005258805.1
XM_017026598.1 Intron XP_016882087.1

View Full Product Details