Product Details

SNP ID
rs10409867
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48900308 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAGTGGGCGTGGTCTAGGGGGGA[A/C]GTCACCAAGAACGCACCGGAGGTCC
Phenotype
MIM: 601323 MIM: 602309
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
NUCB1 PubMed Links

Gene Details

Gene
NUCB1
Gene Name
nucleobindin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006184.5 259 UTR 5 NP_006175.2
XM_017026845.1 259 Intron XP_016882334.1
Gene
TULP2
Gene Name
tubby like protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003323.2 259 Intron NP_003314.2
XM_006723347.3 259 Intron XP_006723410.1
XM_006723348.3 259 Intron XP_006723411.1
XM_011527252.2 259 Intron XP_011525554.1
XM_011527253.2 259 Intron XP_011525555.1
XM_011527254.2 259 Intron XP_011525556.1
XM_011527255.2 259 Intron XP_011525557.1
XM_011527256.2 259 Intron XP_011525558.1

View Full Product Details