Product Details

SNP ID
rs10853787
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47351628 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCATGTTCCAGAGCTACCACAAA[C/G]AGCTCCAGGCTTATATCAAACTTGC
Phenotype
MIM: 609949 MIM: 615475
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C5AR2 PubMed Links

Gene Details

Gene
C5AR2
Gene Name
complement component 5a receptor 2
There are no transcripts associated with this gene.

Gene
DHX34
Gene Name
DEAH-box helicase 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014681.5 204 Intron NP_055496.2
XM_005259443.3 204 Intron XP_005259500.1
XM_011527550.2 204 UTR 5 XP_011525852.1
XM_011527551.2 204 Intron XP_011525853.1
XM_017027524.1 204 Intron XP_016883013.1

View Full Product Details