Product Details

SNP ID
rs11880843
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39973082 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTACATTGTGATTATCTGTATCT[A/G]TCTCTTACTTTTTAGATAGATGTGT
Phenotype
MIM: 602707
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSMC4 PubMed Links

Gene Details

Gene
PSMC4
Gene Name
proteasome 26S subunit, ATPase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006503.3 Intron NP_006494.1
NM_153001.2 Intron NP_694546.1

View Full Product Details